Merck
Pressure-position relations in the glenohumeral joint.
Acta orthopaedica Belgica (1992-01-01)
T Lind, P J Blyme, H H Strange-Vognsen, J Bagger
RÉSUMÉ
The intra-articular pressures in the glenohumeral joint in different positions were measured in 12 cadaveric shoulders. The lowest pressure was found in abduction combined with traction. A significant rise in pressure was seen in positions combining flexion or abduction with rotation. The injected volume to produce a certain pressure as well as the maximum pressure attained in each shoulder showed great variation. These factors are important considerations for immobilization of the shoulder and manipulation during arthroscopy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Sesamolin, ≥97%, from Sesamum indicum (sesame)