Merck
Podocyte specific deletion of PKM2 ameliorates LPS-induced podocyte injury through beta-catenin.
Cell communication and signaling : CCS (2022-06-01)
Mohammed Alquraishi, Samah Chahed, Dina Alani, Dexter L Puckett, Presley D Dowker, Katelin Hubbard, Yi Zhao, Ji Yeon Kim, Laurentia Nodit, Huma Fatima, Dallas Donohoe, Brynn Voy, Winyoo Chowanadisai, Ahmed Bettaieb
RÉSUMÉ
Acute kidney injury (AKI) is associated with a severe decline in kidney function caused by abnormalities within the podocytes' glomerular matrix. Recently, AKI has been linked to alterations in glycolysis and the activity of glycolytic enzymes, including pyruvate kinase M2 (PKM2). However, the contribution of this enzyme to AKI remains largely unexplored. Cre-loxP technology was used to examine the effects of PKM2 specific deletion in podocytes on the activation status of key signaling pathways involved in the pathophysiology of AKI by lipopolysaccharides (LPS). In addition, we used lentiviral shRNA to generate murine podocytes deficient in PKM2 and investigated the molecular mechanisms mediating PKM2 actions in vitro. Specific PKM2 deletion in podocytes ameliorated LPS-induced protein excretion and alleviated LPS-induced alterations in blood urea nitrogen and serum albumin levels. In addition, PKM2 deletion in podocytes alleviated LPS-induced structural and morphological alterations to the tubules and to the brush borders. At the molecular level, PKM2 deficiency in podocytes suppressed LPS-induced inflammation and apoptosis. In vitro, PKM2 knockdown in murine podocytes diminished LPS-induced apoptosis. These effects were concomitant with a reduction in LPS-induced activation of β-catenin and the loss of Wilms' Tumor 1 (WT1) and nephrin. Notably, the overexpression of a constitutively active mutant of β-catenin abolished the protective effect of PKM2 knockdown. Conversely, PKM2 knockdown cells reconstituted with the phosphotyrosine binding-deficient PKM2 mutant (K433E) recapitulated the effect of PKM2 depletion on LPS-induced apoptosis, β-catenin activation, and reduction in WT1 expression. Taken together, our data demonstrates that PKM2 plays a key role in podocyte injury and suggests that targetting PKM2 in podocytes could serve as a promising therapeutic strategy for AKI. Not applicable. Video abstract.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Anti-PKM2 (isoform M1) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody