Merck
  • Measurements of binary diffusion coefficients, retention factors and partial molar volumes for myristoleic acid and its methyl ester in supercritical carbon dioxide.
Measurements of binary diffusion coefficients, retention factors and partial molar volumes for myristoleic acid and its methyl ester in supercritical carbon dioxide.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-11-14)
Chang Yi Kong, Masakazu Mori, Toshitaka Funazukuri, Seiichiro Kagei
RÉSUMÉ
The binary diffusion coefficients, D(12), and retention factors for myristoleic acid and its methyl ester at infinite dilution were measured by the chromatographic impulse response technique in supercritical carbon dioxide at temperatures of 313.2, 333.2 and 343.2 K and pressures from 9.2 to 30 MPa for the acid, and from 8.0 to 14 MPa for the ester. Although the D(12) values were represented by the two correlations, the D(12)/T vs. CO(2) viscosity and the Schmidt-number correlations, which are valid for more than 40 compounds that we have measured so far, significant temperature dependences were observed for the ester. Moreover, the D(12) values for the ester at 313.2 K downward deviated from the background values around 400 kg m(-3), where the partial molar volumes, obtained from the correlation between the retention factors measured and CO(2) densities, showed large negative values.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Myristoléate de méthyle, ≥98.5% (capillary GC), liquid