Merck
  • Molecular docking and molecular dynamics simulation approach to screen natural compounds for inhibition of Xanthomonas oryzae pv. Oryzae by targeting peptide deformylase.
Molecular docking and molecular dynamics simulation approach to screen natural compounds for inhibition of Xanthomonas oryzae pv. Oryzae by targeting peptide deformylase.
Journal of biomolecular structure & dynamics (2020-01-23)
Tushar Joshi, Tanuja Joshi, Priyanka Sharma, Subhash Chandra, Veena Pande
RÉSUMÉ
Xanthomonas oryzae pv. Oryzae (Xoo) causes bacterial leaf blight (BLB) of rice which results in a huge loss in production. Many chemicals are used to control BLB disease. However, these chemicals are toxic to the environments, animals and human beings. Thus, there is a demand to discover potential and safe natural pesticides to manage BLB disease successfully. Therefore, we screened a library of phytochemicals of different plants having antibacterial activity by targeting Peptide Deformylase (PDF) of Xoo using in silico techniques. A library of 318 phytochemicals was prepared and subjected to rigid and flexible molecular docking against PDF followed by molecular dynamics simulation and free energy analysis of protein-ligand complexes. The results of virtual screening showed that 14 compounds from different plants have good binding energy as compare to reference molecule (3 R)-2,3-dihydro[1,3] thiazolo [3,2 a]benzimidazol-3-ol) (-7.7 kcal mol-1). Out of 14 hit compounds, eight compounds that were selected based on binding energy were analyzed by Molecular dynamic (MD) simulation. Analysis of MD simulation revealed that eight compounds namely; Bisdemethoxycurcumin, Rosmarinic acid, Piperanine, Dihydropiperlonguminine, Piperdardine, Dihydrocurcumin and Lonhumosides B achieved good stability during the 80 ns MD simulation at 300 K in term of the RMSD. Further, we calculated RMSF, RG, SASA, and interaction energy after 40 ns due to showing the stability of complexes. From our results, we conclude that these natural compounds could inhibit Xoo by targeting PDF receptor and can be used as potential bactericidal candidates against BLB disease of rice against Xoo and other bacteria. Communicated by Ramaswamy H. Sarma.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Piperanine, ≥95% (LC/MS-ELSD)