Merck
Notch-1 promotes the malignant progression of osteosarcoma through the activation of cell division cycle 20.
Aging (2021-01-08)
Yuan Gao, Lunhao Bai, Guanning Shang
RÉSUMÉ
The molecular mechanism of osteosarcoma (OS) pathogenesis is poorly understood. The Notch signaling pathway has been shown to be critically involved in tumorigenesis, including OS. Therefore, we explored the molecular mechanism by which the Notch-1 signaling pathway is involved in OS progression. Several approaches were carried out to determine the biological function of Notch-1 in OS cells. The MTT results revealed that Notch-1 overexpression increased the viability of OS cells, whereas Notch-1 downregulation reduced cell viability. Consistently, modulation of Notch-1 regulated apoptosis and the migratory and invasive abilities of OS cells. Mechanistic studies showed that Notch-1 overexpression augmented cell division cycle 20 (Cdc20) expression in OS cells. Moreover, overexpression of Cdc20 alleviated the inhibitory effects of Notch-1 downregulation on the viability, migration and invasion of OS cells. Our study offers a promising OS treatment strategy by inhibiting Notch-1.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human CDC20