Merck
Abnormal neocortex arealization and Sotos-like syndrome-associated behavior in Setd2 mutant mice.
Science advances (2021-02-02)
Lichao Xu, Yue Zheng, Xuejing Li, Andi Wang, Dawei Huo, Qinglan Li, Shikang Wang, Zhiyuan Luo, Ying Liu, Fuqiang Xu, Xudong Wu, Min Wu, Yan Zhou
RÉSUMÉ
Proper formation of area identities of the cerebral cortex is crucial for cognitive functions and social behaviors of the brain. It remains largely unknown whether epigenetic mechanisms, including histone methylation, regulate cortical arealization. Here, we removed SETD2, the methyltransferase for histone 3 lysine-36 trimethylation (H3K36me3), in the developing dorsal forebrain in mice and showed that Setd2 is required for proper cortical arealization and the formation of cortico-thalamo-cortical circuits. Moreover, Setd2 conditional knockout mice exhibit defects in social interaction, motor learning, and spatial memory, reminiscent of patients with the Sotos-like syndrome bearing SETD2 mutations. SETD2 maintains the expression of clustered protocadherin (cPcdh) genes in an H3K36me3 methyltransferase-dependent manner. Aberrant cortical arealization was recapitulated in cPcdh heterozygous mice. Together, our study emphasizes epigenetic mechanisms underlying cortical arealization and pathogenesis of the Sotos-like syndrome.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Monoclonal Anti-FOXP2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL5310, purified immunoglobulin, buffered aqueous glycerol solution