Merck
  • Synthesis and biological evaluation of a novel series of pyrazole chalcones as anti-inflammatory, antioxidant and antimicrobial agents.
Synthesis and biological evaluation of a novel series of pyrazole chalcones as anti-inflammatory, antioxidant and antimicrobial agents.
Bioorganic & medicinal chemistry (2009-11-10)
Babasaheb P Bandgar, Shrikant S Gawande, Ragini G Bodade, Nalini M Gawande, Chandrahasya N Khobragade
RÉSUMÉ
A novel series of 1-(2,4-dimethoxy-phenyl)-3-(1,3-diphenyl-1H-pyrazol-4-yl)-propenone (3) have been prepared by the Claisen-Schmidt condensation of 1-(2,4-dimethoxy-phenyl)-ethanone (1) and substituted 1,3-diphenyl-1H-pyrazole-4-carbaldehydes (2). Substituted 1,3-diphenyl-1H-pyrazole-4-carbaldehydes (2) were prepared by Vilsmeir-Haack reaction on acetophenonephenylhydrazones to offer the target compounds. The structures of the compounds were established by IR, (1)H NMR and mass spectral analysis. All the compounds were evaluated for their anti-inflammatory (TNF-alpha and IL-6 inhibitory assays), antioxidant (DPPH free radical scavenging assay) and antimicrobial activities (agar diffusion method) against some pathogenic bacteria and fungi. Of 10 compounds screened, compounds 3a, 3c and 3g exhibited promising IL-6 inhibitory (35-70% inhibition, 10 microM), free radical scavenging (25-35% DPPH activity) and antimicrobial activities (MIC 100 microg/mL and 250 microg/mL) at varied concentrations. The structure-activity relationship (SAR) and in silico drug relevant properties (HBD, HBA, PSA, cLogP, molecular weight, E(HOMO) and E(LUMO)) further confirmed that the compounds are potential lead compounds for future drug discovery study. Toxicity of the compounds was evaluated theoretically and experimentally and revealed to be nontoxic except 3d and 3j.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dexaméthasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexaméthasone, ≥98% (HPLC), powder
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Dexaméthasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexaméthasone, meets USP testing specifications
Sigma-Aldrich
Dexaméthasone, tested according to Ph. Eur.
Supelco
Dexaméthasone, VETRANAL®, analytical standard