Merck
Magnolol inhibits venous remodeling in mice.
Scientific reports (2017-12-21)
Hanna Kuk, Caroline Arnold, Ralph Meyer, Markus Hecker, Thomas Korff
RÉSUMÉ
Due to gravity the venous vasculature in the lower extremities is exposed to elevated pressure levels which may be amplified by obesity or pregnancy. As a consequence, venules dilate and may be slowly transformed into varicose or spider veins. In fact, chronically elevated venous pressure was sufficient to cause the corkscrew-like enlargement of superficial veins in mice. We hypothesized that biomechanical activation of endothelial cells contributes to this process and investigated the inhibitory capacity of Magnolol in this context - a natural compound that features multiple properties counteracting cellular stress. While Magnolol did not influence endothelial capillary sprout formation, it interfered with proliferation, ERK1/2 activity, gelatinase activity as well as baseline production of reactive oxygen species in these cells or murine veins. The anti-oxidative and anti-proliferative capacity of Magnolol was mediated through stimulation of heme oxygenase-1 expression. Finally, local transdermal application of Magnolol attenuated pressure-mediated development of varicose/spider veins in mice and was accompanied by the absence of proliferating and MMP-2 positive endothelial cells. Collectively, our data identified Magnolol as a potent inhibitor of biomechanically evoked endothelial cell activity during pressure-mediated venous remodeling processes which contribute to the development of varicose and spider veins.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Magnolol, ≥90% (HPLC), from plant