Merck
Bioactive Elastic Scaffolds Loaded with Neural Stem Cells Promote Rapid Spinal Cord Regeneration.
ACS biomaterials science & engineering (2021-01-16)
Zhe Gong, Dong Lei, Chenggui Wang, Chao Yu, Kaishun Xia, Jiawei Shu, Liwei Ying, Jiangnan Du, Jingkai Wang, Xianpeng Huang, Licheng Ni, Cong Wang, Jingquan Lin, Fangcai Li, Zhengwei You, Chengzhen Liang
RÉSUMÉ
Despite decades of research, spinal cord injury (SCI) still causes irreparable damage to the human body. Key challenges that hinder the regeneration and extension of neurons following SCI must be overcome, including the overexpressed glial scar formation and strong inflammatory responses in lesion tissue. Transplantation of neural stem cells (NSCs) represents a promising therapeutic method due to its beneficial roles like growth factor secretion and anti-inflammation. However, NSCs usually differentiate into astrocytes, which is considered as one potential limitation of current NSC therapy. Herein, we fabricate an elastic poly(sebacoyl diglyceride) (PSeD) scaffold to mimic the mechanical properties of the natural spinal cord. The PSeD scaffold is coated with poly(sebacoyl diglyceride)-isoleucine-lysine-valine-alanine-valine-serine (PSeD-IKVAVS) to create a bioactive interface. The core point of this topic is divided into two parts. First, PSeD is a bioelastomer and its mechanical properties are similar to those of the natural spinal cord. This feature reduces the direct stimulation to the spinal cord tissue by the elastomer and then reduces the immune response or resistance caused by the host spinal cord tissue. Second, the IKVAVS peptide modifies PSeD to create a bioactive interface to support NSC growth and differentiation. In the in vivo study, the number of CD68-positive macrophages decreased in the PSeD-IKVAVS/NSC group compared to that in the SCI group (20% vs 60%). The low inflammation induced by the scaffold was beneficial to NSCs, resulting in increased locomotor recovery, as indicated by the increased Basso-Beattie-Bresnahan score (5, the average score in the PSeD-IKVAVS/NSC group, vs 2, the average score in the SCI group). Based on the above two characteristics, a PSeD-IKVAVS bioelastomer is fabricated, which provides a beneficial and bioactive microenvironment for NSCs after transplantation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Triéthylamine, analytical standard
Sigma-Aldrich
Sebacoyl chloride, purum, ≥95.0% (GC)