Merck
MAX Functions as a Tumor Suppressor and Rewires Metabolism in Small Cell Lung Cancer.
Cancer cell (2020-05-30)
Arnaud Augert, Haritha Mathsyaraja, Ali H Ibrahim, Brian Freie, Michael J Geuenich, Pei-Feng Cheng, Sydney P Alibeckoff, Nan Wu, Joseph B Hiatt, Ryan Basom, Adi Gazdar, Lucas B Sullivan, Robert N Eisenman, David MacPherson
RÉSUMÉ
Small cell lung cancer (SCLC) is a highly aggressive and lethal neoplasm. To identify candidate tumor suppressors we applied CRISPR/Cas9 gene inactivation screens to a cellular model of early-stage SCLC. Among the top hits was MAX, the obligate heterodimerization partner for MYC family proteins that is mutated in human SCLC. Max deletion increases growth and transformation in cells and dramatically accelerates SCLC progression in an Rb1/Trp53-deleted mouse model. In contrast, deletion of Max abrogates tumorigenesis in MYCL-overexpressing SCLC. Max deletion in SCLC resulted in derepression of metabolic genes involved in serine and one-carbon metabolism. By increasing serine biosynthesis, Max-deleted cells exhibit resistance to serine depletion. Thus, Max loss results in metabolic rewiring and context-specific tumor suppression.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Méthanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acétate d′ammonium, ≥98%
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Tampon Tris-EDTA, 100X concentrate for Northern and Southern blotting
Sigma-Aldrich
Anticorps monoclonal de souris anti-β-actine−peroxydase antibody produced in mouse, clone AC-15, purified from hybridoma cell culture
SAFC
Glycine
Sigma-Aldrich
Anti-Calcitonin Gene Related Peptide antibody produced in rabbit, whole antiserum