Merck
Biogenic Amine Contents and Microbial Characteristics of Cambodian Fermented Foods.
Foods (Basel, Switzerland) (2020-02-23)
Dalin Ly, Sigrid Mayrhofer, Julia-Maria Schmidt, Ulrike Zitz, Konrad J Domig
RÉSUMÉ
Naturally fermented foods are an important part of the typical diet in Cambodia. However, the food safety status of these products has not been widely studied. The aim of this study was, therefore, to provide an overview of the quality of these foods in relation to microbiology and biogenic amines. Additionally, the obtained results were compared to the habits and practices of Cambodians in handling this type of food. A total of 57 fermented foods (42 fishery and 15 vegetable products) were collected from different retail markets in the capital of Cambodia. Pathogenic Salmonella spp., Listeria spp., and Listeria monocytogenes were not detected in 25 g samples. Generally, less than 102 cfu/g of Staphylococcus aureus, Escherichia coli, Pseudomonas spp., Enterobacteriaceae, and molds were present in the fermented foods. Bacillus cereus group members (<102 to 2.3 × 104 cfu/g), lactic acid bacteria (<102 to 1.1 × 107 cfu/g), halophilic and halotolerant bacteria (<102 to 8.9 × 106 cfu/g), sulfite-reducing Clostridium spp. (<102 to 3.5 × 106 cfu/g), and yeasts (<102 to 1.1 × 106 cfu/g) were detected in this study. Still, the presence of pathogenic and spoilage microorganisms in these fermented foods was within the acceptable ranges. Putrescine, cadaverine, tyramine, and histamine were detected in 100%, 89%, 81%, and 75% of the tested products, respectively. The concentrations of histamine (>500 ppm) and tyramine (>600 ppm) were higher than the recommended maximum levels in respectively four and one of 57 fermented foods, which represents a potential health risk. The results suggest that the production process, distribution, and domestic handling of fermented foods should be re-evaluated. Further research is needed for the establishment of applicable preservation techniques in Cambodia.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Heptylamine, 99%