Merck
Preparation, characterization and degradation study of novel sulfonated furanic poly(ester-amide)s.
Designed monomers and polymers (2020-03-05)
Majdi Abid, Sirine Mhiri, Abdelkader Bougarech, Rania Triki, Souhir Abid
RÉSUMÉ
A series of novel-sulfonated furanic polyesteramides have been synthesized by polycondensation in the melt-phase. The copolymers were obtained with reasonable molecular weights and high inherent viscosities between 0.2 and 0.4 dL/g. The hydrolytic degradability of polyesteramides was enhanced by the increasing of amide units in the copolymers backbone.