Merck
Effect of L-carnitine and palmitoyl-L-carnitine on erythroid colony formation in fetal mouse liver cell culture.
American journal of nephrology (1998-09-08)
M Matsumura, S Hatakeyama, I Koni, H Mabuchi
RÉSUMÉ
The administration of L-carnitine to patients undergoing hemodialysis increases hematocrit and improves the response to recombinant human erythropoietin (rhEPO). This suggests a contribution by carnitine to erythrocyte membrane function or erythropoiesis. We investigated the effect of L-carnitine and palmitoyl-L-carnitine on erythropoiesis by assessing erythroid colony formation in in vitro fetal mouse liver cell cultures. L-Carnitine or palmitoyl-L-carnitine was added with rhEPO to fetal mouse liver cell cultures. Doses of L-carnitine of up to 200 micromol/l to the culture had no effect on colony formation. In contrast, the addition of above 12.5 micromol/l palmitoyl-L-carnitine into the culture increased colony formation significantly. These results suggest that long-chain acyl carnitine may have an effect on erythropoiesis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Palmitoyl-L-carnitine chloride, ≥98% (TLC), powder
Sigma-Aldrich
Palmitoyl-DL-carnitine chloride, powder