Merck
  • Double diastereoselection in aldol reactions mediated by dicyclohexylchloroborane between chiral aldehydes and a chiral ethyl ketone derived from L-erythrulose. synthesis of a C1-C9 fragment of the structure of the antifungal metabolite soraphen A1alpha.
Double diastereoselection in aldol reactions mediated by dicyclohexylchloroborane between chiral aldehydes and a chiral ethyl ketone derived from L-erythrulose. synthesis of a C1-C9 fragment of the structure of the antifungal metabolite soraphen A1alpha.
The Journal of organic chemistry (2005-11-10)
Santiago Díaz-Oltra, Juan Murga, Eva Falomir, Miguel Carda, Gabriel Peris, J Alberto Marco
RÉSUMÉ
[Chemical reaction: See text] Both matched and mismatched diastereoselections have been observed in the aldol reactions of a range of chiral aldehydes with the dicyclohexylboron enolate of a chiral ethyl ketone related to L-erythrulose. As was previously observed in the corresponding aldol reactions with L-erythrulose derivatives, the Felkin-Anh model provides an adequate explanation for the stereochemical outcome of reactions with chiral alpha-methyl aldehydes. However, a satisfactory account of the results observed with alpha-oxygenated aldehydes was only possible with the Cornforth model. As a practical application of the methodology described herein, a C1-C9 fragment of the structure of the antifungal macrolide soraphen A1alpha has been prepared in a convergent and stereoselective way.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-(+)-Erythrulose, ≥75% (HPLC)