Merck
  • Microbial Communities Associated with Sustained Anaerobic Reductive Dechlorination of α-, β-, γ-, and δ-Hexachlorocyclohexane Isomers to Monochlorobenzene and Benzene.
Microbial Communities Associated with Sustained Anaerobic Reductive Dechlorination of α-, β-, γ-, and δ-Hexachlorocyclohexane Isomers to Monochlorobenzene and Benzene.
Environmental science & technology (2019-12-14)
Wenjing Qiao, Luz A Puentes Jácome, Xianjin Tang, Line Lomheim, Minqing Ivy Yang, Sarra Gaspard, Ingrid Regina Avanzi, Jichun Wu, Shujun Ye, Elizabeth A Edwards
RÉSUMÉ
Intensive historical and worldwide use of pesticide formulations containing hexachlorocyclohexane (HCH) has led to widespread contamination. We derived four anaerobic enrichment cultures from HCH-contaminated soil capable of sustainably dechlorinating each of α-, β-, γ-, and δ-HCH isomers stoichiometrically to benzene and monochlorobenzene (MCB). For each isomer, the dechlorination rates, inferred from production rates of the dechlorinated products, MCB and benzene, increased progressively from <3 to ∼12 μM/day over 2 years. The molar ratio of benzene to MCB produced was a function of the substrate isomer and ranged from β (0.77 ± 0.15), α (0.55 ± 0.09), γ (0.13 ± 0.02), to δ (0.06 ± 0.02) in accordance with pathway predictions based on prevalence of antiperiplanar geometry. Data from 16S rRNA gene amplicon sequencing and quantitative PCR revealed significant increases in the absolute abundances of Pelobacter and Dehalobacter, most notably in the α-HCH and δ-HCH cultures. Cultivation with a different HCH isomer resulted in distinct bacterial communities, but similar archaeal communities. This study provides the first direct comparison of shifts in anaerobic microbial communities induced by the dechlorination of distinct HCH isomers. It also uncovers candidate microorganisms responsible for the dechlorination of α-, β-, γ-, and δ-HCH, a key step toward better understanding and monitoring of natural attenuation processes and improving bioremediation technologies for HCH-contaminated sites.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
HCH, PESTANAL®, analytical standard, mixture of isomers (α:β:γ:δ=1:1:1:1)