Merck
Amyloid beta-peptide Abeta(1-42) but not Abeta(1-40) attenuates synaptic AMPA receptor function.
Synapse (New York, N.Y.) (2007-03-21)
Kodeeswaran Parameshwaran, Catrina Sims, Patrick Kanju, Thirumalini Vaithianathan, Brian C Shonesy, Muralikrishnan Dhanasekaran, Ben A Bahr, Vishnu Suppiramaniam
RÉSUMÉ
The brains of Alzheimer's disease (AD) patients have large numbers of plaques that contain amyloid beta (Abeta) peptides which are believed to play a pivotal role in AD pathology. Several lines of evidence have established the inhibitory role of Abeta peptides on hippocampal memory encoding, a process that relies heavily on alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) receptor function. In this study the modulatory effects of the two major Abeta peptides, Abeta(1-40) and Abeta(1-42), on synaptic AMPA receptor function was investigated utilizing the whole cell patch clamp technique and analyses of single channel properties of synaptic AMPA receptors. Bath application of Abeta(1-42) but not Abeta(1-40) reduced both the amplitude and frequency of AMPA receptor mediated excitatory postsynaptic currents in hippocampal CA1 pyramidal neurons by approximately 60% and approximately 45%, respectively, in hippocampal CA1 pyramidal neurons. Furthermore, experiments with single synaptic AMPA receptors reconstituted in artificial lipid bilayers showed that Abeta(1-42) reduced the channel open probability by approximately 42% and channel open time by approximately 65% and increased the close times by several fold. Abeta(1-40), however, did not show such inhibitory effects on single channel properties. Application of the reverse sequence peptide Abeta(42-1) also did not alter the mEPSC or single channel properties. These results suggest that Abeta(1-42) but not Abeta(1-40) closely interacts with and exhibits inhibitory effects on synaptic AMPA receptors and may contribute to the memory impairment observed in AD.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Amyloid β 42-1 reverse human, ≥95% (HPLC)