Merck
Comparative Functional Analysis of 12 Mammalian IFN-λ4 Orthologs.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research (2015-08-27)
Ashley Paquin, Olusegun O Onabajo, Wei Tang, Ludmila Prokunina-Olsson
RÉSUMÉ
IFN-λ4 is a novel type-III interferon with strong clinical significance in humans. Only a subset of individuals--up to 10% of Asians, 50% of Europeans, and 90% of Africans--carry the ΔG allele of a genetic variant rs368234815-TT/ΔG and are genetically able to produce IFN-λ4 protein. Carriers of the ΔG allele have impaired ability to clear infection with hepatitis C virus (HCV). IFN-λ4 is also predicted to exist and be functionally important in several nonhuman mammals. In this study, we present the first comparative analysis of 12 mammalian IFN-λ4 orthologs in a human hepatic cell line, HepG2, which supports signaling of the human IFN-λ4. We show that despite differences in protein sequences, functional properties of the recombinant human and nonhuman IFN-λ4 proteins are comparable-they are all expressed as predominantly cytoplasmic proteins that are biologically active for induction of interferon signaling. We show that several IFN-λ4 orthologs can be detected by Western blotting, flow cytometry, and confocal imaging using a monoclonal antibody developed for the human IFN-λ4. Studies of IFN-λ4 in animals should help improve our understanding of the biology of this novel clinically important interferon in normal and disease conditions.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-Interferon Lambda 4, clone 4G1, clone 4G1, 1 mg/mL, from mouse