Merck
  • Fibroblasts from Werner syndrome patients: cancer cells derived by experimental introduction of oncogenes maintain malignant properties despite entering crisis.
Fibroblasts from Werner syndrome patients: cancer cells derived by experimental introduction of oncogenes maintain malignant properties despite entering crisis.
Oncology reports (2010-01-01)
Furong Yuan, Meizhen Chen, Peter J Hornsby
RÉSUMÉ
Werner syndrome (WS) results from defects in the gene encoding WRN RecQ helicase. WS fibroblasts undergo premature senescence in culture. Because cellular senescence is a tumor suppressor mechanism, we examined whether WS fibroblasts exhibited reduced tumorigenicity, in comparison to control cells, in a model of experimental conversion of normal human cells to cancer cells. The combination of oncogenic Ras (Ha-Ras(V12G)) and SV40 large T antigen (SV40 LT) causes human cells to acquire neoplastic properties in the absence of telomerase. We found that WS cells could also be converted to a tumorigenic state by these oncogenes, as evidenced by invasion and metastasis of cells implanted in immunodeficient mice. Ras/SV40 LT-expressing cells retained invasiveness and malignant properties even when cells reached crisis in tumors in vivo. High levels of gelatinase were found by an in situ assay in Ras/SV40 LT-expressing cells undergoing crisis. We conclude that, despite evidence of accelerated senescence in WS cells, there is no evidence that the absence of active WRN acts as a barrier to neoplastic transformation. Moreover, we find that tumorigenic human cells retain malignant properties of the cells as they approach and reach crisis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-grand antigène T du virus SV40, clone PAb416, clone PAb416, from mouse