Merck
Intestinal Drug Interactions Mediated by OATPs: A Systematic Review of Preclinical and Clinical Findings.
Journal of pharmaceutical sciences (2017-04-18)
Jingjing Yu, Zhu Zhou, Jessica Tay-Sontheimer, René H Levy, Isabelle Ragueneau-Majlessi
RÉSUMÉ
In recent years, an increasing number of clinical drug-drug interactions (DDIs) have been attributed to inhibition of intestinal organic anion-transporting polypeptides (OATPs); however, only a few of these DDI results were reflected in drug labels. This review aims to provide a thorough analysis of intestinal OATP-mediated pharmacokinetic-based DDIs, using both in vitro and clinical investigations, highlighting the main mechanistic findings and discussing their clinical relevance. On the basis of pharmacogenetic and clinical DDI results, a total of 12 drugs were identified as possible clinical substrates of OATP2B1 and OATP1A2. Among them, 3 drugs, namely atenolol, celiprolol, and fexofenadine, have emerged as the most sensitive substrates to evaluate clinical OATP-mediated intestinal DDIs when interactions with P-glycoprotein by the test compound can be ruled out. With regard to perpetrators, 8 dietary or natural products and 1 investigational drug, ronacaleret (now terminated), showed clinical intestinal inhibition attributable to OATPs, producing ≥20% decreases in area under the plasma concentration-time curve of the co-administered drug. Common juices, such as apple juice, grapefruit juice, and orange juice, are considered potent inhibitors of intestinal OATP2B1 and OATP1A2 (decreasing exposure of the co-administered substrate by ∼85%) and may be adequate prototype inhibitors to investigate intestinal DDIs mediated by OATPs.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Celiprolol hydrochloride, ≥98% (HPLC)