Merck
A functional connectome phenotyping dataset including cognitive state and personality measures.
Scientific data (2019-02-13)
Natacha Mendes, Sabine Oligschläger, Mark E Lauckner, Johannes Golchert, Julia M Huntenburg, Marcel Falkiewicz, Melissa Ellamil, Sarah Krause, Blazej M Baczkowski, Roberto Cozatl, Anastasia Osoianu, Deniz Kumral, Jared Pool, Laura Golz, Maria Dreyer, Philipp Haueis, Rebecca Jost, Yelyzaveta Kramarenko, Haakon Engen, Katharina Ohrnberger, Krzysztof J Gorgolewski, Nicolas Farrugia, Anahit Babayan, Andrea Reiter, H Lina Schaare, Janis Reinelt, Josefin Röbbig, Marie Uhlig, Miray Erbey, Michael Gaebler, Jonathan Smallwood, Arno Villringer, Daniel S Margulies
RÉSUMÉ
The dataset enables exploration of higher-order cognitive faculties, self-generated mental experience, and personality features in relation to the intrinsic functional architecture of the brain. We provide multimodal magnetic resonance imaging (MRI) data and a broad set of state and trait phenotypic assessments: mind-wandering, personality traits, and cognitive abilities. Specifically, 194 healthy participants (between 20 and 75 years of age) filled out 31 questionnaires, performed 7 tasks, and reported 4 probes of in-scanner mind-wandering. The scanning session included four 15.5-min resting-state functional MRI runs using a multiband EPI sequence and a hig h-resolution structural scan using a 3D MP2RAGE sequence. This dataset constitutes one part of the MPI-Leipzig Mind-Brain-Body database.