Merck
Degradation of Dicloran in Irradiated Water-Sediment Systems.
Journal of agricultural and food chemistry (2019-07-02)
Emily N Vebrosky, Laura M Basirico, Kevin L Armbrust
RÉSUMÉ
Shallow water systems are uniquely susceptible to environmental processes such as photolysis and hydrolysis that can influence the dissipation of pesticides into sediments. The fungicide dicloran has previously been shown to undergo photolysis and is reported to dissipate in soils and sediments. The photodegradation and dissipation of dicloran in freshwater and seawater was monitored in a laboratory-simulated shallow water system. While no difference was observed between freshwater and seawater systems in the presence of simulated sunlight, the dissipation of dicloran in dark trial systems differed between salinities; 30% of the applied mass dissipated into the sediment in freshwater vs 22% in seawater, and the photodegradation rate and half-life were also impacted by the presence of sediment. The potential for dicloran to dissipate and photodegrade affects the overall behavior of dicloran between waters. Differences in chemical behavior with sediment presence and potential for photodegradation have the capacity to impact organisms within the ecosystem and suggest that these factors may need to be implemented into chemical exposure assessments dependent upon location.