Merck
  • Monitoring of Air-Dispersed Formaldehyde and Carbonyl Compounds as Vapors and Adsorbed on Particulate Matter by Denuder-Filter Sampling and Gas Chromatographic Analysis.
Monitoring of Air-Dispersed Formaldehyde and Carbonyl Compounds as Vapors and Adsorbed on Particulate Matter by Denuder-Filter Sampling and Gas Chromatographic Analysis.
International journal of environmental research and public health (2019-06-06)
Stefano Dugheri, Nicola Mucci, Giovanni Cappelli, Alessandro Bonari, Giacomo Garzaro, Giorgio Marrubini, Gianluca Bartolucci, Marcello Campagna, Giulio Arcangeli
RÉSUMÉ
Carbonyl compounds (CCs) are products present both as vapors and as condensed species adsorbed on the carbonaceous particle matter dispersed in the air of urban areas, due to vehicular traffic and human activities. Chronic exposure to CCs is a potential health risk given the toxicity of these chemicals. The present study reports on the measurement of the concentrations of 14 CCs in air as vapors and 2.5 µm fraction PM by the ENVINT GAS08/16 gas/aerosol sampler, a serial sampler that uses annular denuder, as sampling device. The 14 CCs were derivatized during sampling prior to gas-chromatographic separation and multiple detection by mass spectrometry, nitrogen-phosphorus thermionic, electron capture detection. Outdoor air multiple samples were collected in four locations in the urban area of Florence. The results evidenced that formaldehyde, acetaldehyde, and acetone were the more abundant CCs in the studied areas. The data collected was discussed considering the particle to vapor ratio of each CC found. The CCs pollution picture obtained was tentatively related to the nature and intensity of the traffic transiting by the sampling sites. This approach allowed to determine 14 CCs in both concentrated and diluted samples and is proposed as a tool for investigating outdoor and indoor pollution.