Merck
  • Estimates of calf starter energy affected by consumption of nutrients. 2. Effect of changing digestion on energy content in calf starters.
Estimates of calf starter energy affected by consumption of nutrients. 2. Effect of changing digestion on energy content in calf starters.
Journal of dairy science (2019-01-30)
J D Quigley, W Hu, J R Knapp, T S Dennis, F X Suarez-Mena, T M Hill
RÉSUMÉ
Apparent total-tract digestibility data from 3 published studies with calves from 0 to 4 mo of age were used to evaluate National Research Council (NRC; 2001) estimates of digestible energy (DE) and metabolizable energy (ME) in calf starters (CS). Calves (n = 83) or pens of calves (n = 24) were used in model development. In each study, 48 Holstein bull calves (2-3 d of age at initiation of each study) were fed varying amounts of milk replacer with CS and water for ad libitum consumption. Calf starters varied in nutrient composition and physical form (pelleted, textured, or mixed with 5% grass hay and fed as a total mixed ration). Apparent total-tract digestibility was measured at various ages. Feed and feces were collected from 20 calves per trial during 5-d collection periods during the first 56 d of each trial. In 2 studies, calves were grouped in pens (4 calves/pen) for a second 56-d measurement period. Fecal collections were repeated occasionally during the second period. Total-tract digestibilities (n = 207) of neutral detergent fiber, nonfiber carbohydrates (NFC), crude protein (CP), and fat were used to calculate ME in CS using equations from the 2001 Dairy NRC. Contributions of digestible CP and fat from milk replacer before weaning were estimated using nonlinear regression and removed from estimates of fat and CP digestibility in CS. Digestion of most nutrients in CS and calculated DE and ME in CS were low early in life and increased with increasing cumulative NFC intake. The natural logarithm of cumulative NFC intake, measured from d 0 to the end of each digestibility period, accounted for more variation in CS nutrient digestibilities, DE and ME estimates compared with daily NFC intake or intake of other nutrients, intake of milk replacer, or age of calf. Calculated ME values in CS were similar to those predicted by NRC after calves consumed approximately 15 kg of cumulative NFC or 28 kg of cumulative dry matter intake (assuming 53% NFC in CS). Current estimates of energy in CS fed to 4 mo of age may overestimate contribution of dry feed to overall energy metabolism in young calves.