Merck
Involvement of the HIV-1 external envelope glycoprotein 120 (gp120) C2 region in gp120 oligomerization.
Biochimica et biophysica acta (1997-07-18)
N Seddiki, H Bouhlal, L Rabehi, A Benjouad, C Devaux, J C Gluckman, L Gattegno
RÉSUMÉ
A synthetic peptide resembling the C2 region of human immunodeficiency virus type 1 (HIV-1) gp120 (C2-Lai: amino acids (aa) 273-288), inhibited (C50 = 200 microM) gp120 calcium-dependent binding of N-acetyl-beta-D-glucosaminyl and mannosyl residues exposed on natural glycoprotein bovine fetuin whereas a peptide derived from an aa sequence downstream of C2-Lai (C2-SC19) had no such effect (C50 > 1000 microM). No calcium-carbohydrate-specific binding of C2-Lai to fetuin was detected. In addition, C2-Lai was also found to inhibit the calcium-dependent oligomerization of gp120: while recombinant gp120 (rgp120) was recovered mainly as oligomers (78%) in 10 mM CaCl2, in contrast to 100% monomers in 1mM CaCl2, mostly monomers (67%) were found in 10 mM CaCl2 in the presence of C2-Lai. Peptide C2-SC19 and carbohydrate structures such as fetuin, fucoidin, dextran or mannan did not significantly affect gp120 oligomerization. Electrophoresis and gel filtration analysis also showed that C2-Lai aggregated in the form of 20 kDa compounds, which is compatible with association of 10 molecules. Taken together, these results demonstrate that the C2 domain is involved in gp120 oligomerization and suggest that gp120 oligomers but not monomers have specific carbohydrate binding properties.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Fetuin-Agarose, saline suspension