Merck
Distribution and identity of neurons expressing the oxytocin receptor in the mouse spinal cord.
Neuroscience letters (2011-03-23)
Ludovic Wrobel, Ara Schorscher-Petcu, Anouk Dupré, Masahide Yoshida, Katsuhiko Nishimori, Eliane Tribollet
RÉSUMÉ
Oxytocin can influence various spinal functions. However, little is known about the spinal neuronal networks responsible for oxytocin effects. The aim of this study was to localize and characterize spinal neurons expressing oxytocin receptors. We used an oxytocin receptor-reporter mouse in which the fluorescent protein Venus is expressed under the control of the oxytocin receptor gene promoter. At all segmental levels, Venus-expressing neurons were most numerous in the substantia gelatinosa, mingled with protein kinase Cγ interneurons in the innermost layer of the inner lamina II, which, in contrast to the outer two thirds of this layer, does not receive nociceptive input. Venus-expressing neurons were also observed in the intermediolateral and sacral parasympathetic nuclei, where they represented about 5% of presumed preganglionic neurons identified by choline acetyltransferase immunoreactivity. Finally, Venus immunoreactivity was detected in lumbar and sacral dorsal gray commissures as well as in isolated neurons scattered in different regions of the dorsal horn. Altogether, our results establish the location of neurons putatively involved in oxytocin modulation of spinal functions, in particular of sexual functioning and nociception.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), biotin conjugate, lyophilized powder