Principaux documents
Z685437
Vacuubrand chemistry pumping units, manual control, Model PC 201 NT
AC/DC input 230 V AC, European 2-pin plug
Synonyme(s) :
Vacuubrand 20736403, diaphragm pump
1 EA
5 550.00 CHF
5 550.00 CHF
Date d'expédition estimée le23 avril 2025
Changer de vue
1 EA
5 550.00 CHF
About This Item
Code UNSPSC :
41105101
Nomenclature NACRES :
NB.79
5 550.00 CHF
Date d'expédition estimée le23 avril 2025
Fabricant/nom de marque
Vacuubrand 737000
Paramètres
105 mbar vacuum
3.4-3.8 m3/h pumping speed
Description générale
This chemistry pumping unit has a wide range of applications like evacuation, evaporation and pumping of gases and vapors in chemical, biological and pharmaceutical laboratories. Typical applications are rotary evaporators, vacuum concentrators and vacuum drying ovens with high boiling solvents.
The manual flow control valve regulates the effective pumping speed at the vacuum connection; the vacuum manometer offers an analog vacuum display. The separator at the inlet (AK), made of glass with a protective coating, retains particles and liquid droplets. The exhaust waste vapor condenser enables near-100-percent solvent recovery.
  • Outstanding chemical resistance and superior vapor tolerance
  • Exceptionally high performance even at low vacuum
  • Excellent ultimate vacuum even with gas ballast
  • Manual vacuum control, analog vacuum display
  • Excellent environmental friendliness due to efficient solvent recovery
Supplied with MD 4C, manual flow control valve, analogue vacuum gauge, inlet catchpot and exhaust waste vapor condenser certification (NRTL).