Merck
HomeCell SignalingSodium Channels
Sodium Channels
Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials. A variety of toxins and chemicals are known to either block or modulate sodium channels and have proven invaluable in investigating the physiological characteristics of these channels. Most notably, tetrodotoxin (TTX), isolated from puffer fish, is a potent and selective blocker of sodium channels. Saxitoxin (STX), a toxin isolated from dinoflagellates, was also found to have the same blocking action as TTX. The sodium channels present in brain, along with those found in peripheral nerves and skeletal muscle, are highly sensitive to TTX/STX at nanomolar concentrations, whereas some sodium channels in the heart are blocked at concentrations in the micromolar range, and TTX-resistant sodium channels in dorsal root ganglion neurons are only blocked at concentrations approaching 100 mM.
Structurally, the sodium channels of brain comprise one α subunit of 260 kDa, one β1 subunit of 36 kDa, and one β2 subunit of 33 kDa, forming a heterotrimeric structure. β3 may substitute for β1 and β4 for β2 in these brain sodium channel complexes. The sodium channels of skeletal muscle consist of one α and one β1-like subunit, whereas those of the heart and peripheral neurons are likely to be complexes of α, β and possibly other proteins/subunits, in unknown stoichiometry.
A variety of toxins and chemicals that modulate the function of sodium channels have been discovered and many of these are now being used as chemical tools to study these ion channels. These may be classified into several groups based on the mechanisms of modification of channel gating kinetics and on the binding sites. TTX, STX and µ-conotoxins bind to neurotoxin receptor site 1 to block sodium channels. Batrachotoxin, grayanotoxins, veratridine and aconitine alter the kinetics and voltage dependence of sodium channel activation and inhibit sodium channel inactivation through binding to neurotoxin receptor site 2. The combination of effects of these toxins causes persistent activation of sodium channels at the resting membrane potential. Sea anemone toxins and α-scorpion toxins block the sodium channel inactivation by binding to neurotoxin receptor site 3. The protease pronase and the chemical reagents N-bromo-acetamide and chloramine-T also block inactivation, but their exact sites of action are unknown. α-scorpion toxins shift the voltage dependence of activation negatively by binding to neurotoxin receptor site 4. Brevetoxins and ciguatoxins are similar to batrachotoxin and other site 1 toxins in their alteration of the kinetics and voltage dependence of activation and inhibition of inactivation, but brevetoxins and ciguatoxins bind to neurotoxin receptor site 5. The pyrethroid insecticides alter the voltage dependence and kinetics of both activation and inactivation of sodium channels and appear to bind to neurotoxin receptor site 6.
Sensory ganglion neurons (e.g. dorsal root and nodose ganglia), particularly those associated with small-diameter afferent fibers, express sodium channels that are highly resistant to TTX and play an important role in nociceptive mechanisms. Recent research efforts have focused on the development of agents selective for the TTX-resistant sodium channels of dorsal root ganglia, in anticipation that these might prove to be novel analgesic drugs.
Numerous clinically used drugs block sodium channels. Local anesthetics used in control of acute pain, antiarrhythmic drugs used in therapy of cardiac arrhythmias, and some antiepileptics used in control of seizures all bind to a common local anesthetic receptor site on sodium channels. Many of these drugs exhibit use-dependent block, a characteristic crucial for their therapeutic effects, as the channel block becomes more potent during rapid firing in arrhythmic or epileptic conditions.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
NomenclatureaNaV1.1NaV1.2NaV1.2aNaV1.3NaV1.4
Previous NamesIIIIIAIIIμ1
Primary SourceCNSCNS (neonatal)CNS (adult)CNS (embryonic)Skeletal muscle
Structural Information2009 aa (rat)2005 aa (rat)2005 aa (rat)1951 aa (rat)1840 aa (rat)
Subunitsα, β1-β4α, β1-β4α, β1-β4α, β1-β4α, β1
Conductance 20-25 pS20-25 pS 20-25 pS
Ionic SelectivityNa+ > K+ > Ca2+Na+ > K+ > Ca2+Na+ > K+ > Ca2+Na+ > K+ > Ca2+Na+ > K+ > Ca2+
Blockers 
Neurotoxin Receptor Site 1 
TTX10 nM10 nM10 nM10 nM10 nM
STX2 nM2 nM2 nM2 nM2 nM
μ-ConotoxinN/AN/AN/AN/AActive
Local Anesthetic Receptor SiteLocal Anesthetics (L5647P9879)b
Class I Antiarrhythmic Drugs
Phenytoin (D4505)
Carbamazepine (C4024)
Lamotrigine (L3791)
Modulators 
Neurotoxin Receptor Site 2 (Enhance activation and block inactivation)Batrachotoxin
Grayanotoxin
Veratridine (V5754)
Aconitine (A8001)
Neurotoxin Receptor Site 3 (Slow inactivation)α-Scorpion toxins
Sea anemone toxins (A7475)
Neurotoxin Receptor Site 4 (Enhance activation)β-Scorpion toxins
Neurotoxin Receptor Site 5 (Enhance activation and block inactivation)Brevetoxins
Ciguatoxin
Versutoxin
Neurotoxin Receptor Site 6 (Enhance activation and block inactivation)Pyrethoid insecticides
Radioligands[3H]-Brevetoxin
[3H]-Batrachotoxinin A 20 α-benzoate
[3H]-Saxitoxin
Primary Tissue ExpressionCNSCNSCNSCNSSkeletal muscle
Physiological FunctionAction potential initiation and conduction, synaptic integrationAction potential initiation and conduction, synaptic integrationAction potential initiation and conduction, synaptic integrationAction potential initiation and conduction, synaptic integrationAction potential initiation and conduction, synaptic integration
Disease RelevanceEpilepsy, chronic painEpilepsyEpilepsyEpilepsy, chronic painPeriodic paralysis, paralysis
NomenclatureaNaV1.5NaV1.6NaV1.7NaV1.8NaV1.9
Previous Namesh1VIPN1PN3/SNSSNS2
Primary SourceHeartCNSSympathetic and dorsal root gangliaDorsal root gangliaDorsal root ganglia
Structural Information2019 aa (rat)1976 aa (rat)1984 aa (rat)1957 aa (rat)1765 aa (rat)
Subunitsα, β1-β4α, βα, βα, βα, β
Conductance20-25 pS    
Ionic SelectivityNa+ > K+ > Ca2+Na+ > K+ > Ca2+Na+ > K+ > Ca2+Na+ > K+ > Ca2+Na+ > K+ > Ca2+
Blockers 
Neurotoxin Receptor Site 1 
TTX1 μM10 nM10 nM≥ 50 mM≥ 50 mM
STXActiveActiveActiveNot knownNot known
μ-ConotoxinN/AN/AN/AN/AN/A
Local Anesthetic Receptor SiteLocal Anesthetics (L5647P9879)b
Class I Antiarrhythmic Drugs
Phenytoin (D4505)
Carbamazepine (C4024)
Lamotrigine (L3791)
N/AN/A
Modulators 
Neurotoxin Receptor Site 2 (Enhance activation and block inactivation)Batrachotoxin
Grayanotoxin
Veratridine (V5754)
Aconitine (A8001)
N/AN/A
Neurotoxin Receptor Site 3
(Slow inactivation)
α-Scorpion toxins
Sea anemone toxins (A7475)
N/AN/A
Neurotoxin Receptor Site 4 (Enhance activation)β-Scorpion toxinsN/AN/A
Neurotoxin Receptor Site 5 (Enhance activation and block inactivation)Brevetoxins
Ciguatoxin
Versutoxin
N/AN/A
Neurotoxin Receptor Site 6 (Enhance activation and block inactivation)Pyrethoid insecticidesN/AN/A
Radioligands[3H]-Brevetoxin
[3H]-Batrachotoxinin A 20 α-benzoate
[3H]-Saxitoxin
N/AN/A
Primary Tissue
Expression
HeartCNSPNSDRGDRG
Physiological FunctionAction potential initiation and conductionAction potential initiation and conduction, synaptic integration
Action potential initiation and conduction
Action potential initiation and conductionAction potential initiation and conduction
Disease RelevanceArrhythmiaPainChronic painChronic painChronic pain
Footnotes
a) This nomenclature, proposed by Goldin et al. in Neuron, 28, 365-368 (2000) has been reviewed and accepted by the Nomenclature Committee of IUPHAR.
b) Commonly used local anesthetics include Lidocaine (L5647), Mexiletine (M2727), Procainamide (P9391), Procaine (P9879), Flecainide (F6777) and Tetracaine (T7508). Typically, these local anesthetic, antiarrhythmic, and antiepileptic drugs have similar affinity for NaV1.1-1.7 and reduced affinity for NaV1.8 and NaV1.9.
Abbreviations
CNS: Central nervous system
DRG: Dorsal root ganglion
PNS: Peripheral nervous system
STX: Saxitoxin
TTX: Tetrodotoxin
Similar Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Akopian AN, Sivilotti L, Wood JN. 1996. A tetrodotoxin-resistant voltage-gated sodium channel expressed by sensory neurons. Nature. 379(6562):257-262. https://doi.org/10.1038/379257a0
2.
Brackenbury WJ. 2012. Voltage-gated sodium channels and metastatic disease. Channels. 6(5):352-361. https://doi.org/10.4161/chan.21910
3.
Catterall WA. 2000. From Ionic Currents to Molecular Mechanisms. Neuron. 26(1):13-25. https://doi.org/10.1016/s0896-6273(00)81133-2
4.
Cestèle S. 2000. Molecular mechanisms of neurotoxin action on voltage-gated sodium channels. 82(9-10):883-892. https://doi.org/10.1016/s0300-9084(00)01174-3
5.
Cummins TR, Dib-Hajj SD, Black JA, Akopian AN, Wood JN, Waxman SG. 1999. A Novel Persistent Tetrodotoxin-Resistant Sodium Current In SNS-Null And Wild-Type Small Primary Sensory Neurons. J. Neurosci.. 19(24):RC43-RC43. https://doi.org/10.1523/jneurosci.19-24-j0001.1999
6.
Dib-Hajj SD, Tyrrell L, Black JA, Waxman SG. 1998. NaN, a novel voltage-gated Na channel, is expressed preferentially in peripheral sensory neurons and down-regulated after axotomy. Proceedings of the National Academy of Sciences. 95(15):8963-8968. https://doi.org/10.1073/pnas.95.15.8963
7.
Dib-Hajj SD, Yang Y, Black JA, Waxman SG. 2013. The NaV1.7 sodium channel: from molecule to man. Nat Rev Neurosci. 14(1):49-62. https://doi.org/10.1038/nrn3404
8.
Goldin AL, Barchi RL, Caldwell JH, Hofmann F, Howe JR, Hunter JC, Kallen RG, Mandel G, Meisler MH, Netter YB, et al. 2000. Nomenclature of Voltage-Gated Sodium Channels. Neuron. 28(2):365-368. https://doi.org/10.1016/s0896-6273(00)00116-1
9.
Goldin AL. 2001. Resurgence of Sodium Channel Research. Annu. Rev. Physiol.. 63(1):871-894. https://doi.org/10.1146/annurev.physiol.63.1.871
10.
Narahashi T. 1988. Handbook of Natural Toxins, Marine toxins and Venoms. 3. New York: Marcell Dekker, Inc..
11.
Roberson D, Binshtok A, Blasl F, Bean B, Woolf C. 2011. Targeting of sodium channel blockers into nociceptors to produce long-duration analgesia: a systematic study and review. 164(1):48-58. https://doi.org/10.1111/j.1476-5381.2011.01391.x
12.
Sangameswaran L, Fish LM, Koch BD, Rabert DK, Delgado SG, Ilnicka M, Jakeman LB, Novakovic S, Wong K, Sze P, et al. 1997. A Novel Tetrodotoxin-sensitive, Voltage-gated Sodium Channel Expressed in Rat and Human Dorsal Root Ganglia. J. Biol. Chem.. 272(23):14805-14809. https://doi.org/10.1074/jbc.272.23.14805
13.
Schaller K, Krzemien D, Yarowsky P, Krueger B, Caldwell J. 1995. A novel, abundant sodium channel expressed in neurons and glia. J. Neurosci.. 15(5):3231-3242. https://doi.org/10.1523/jneurosci.15-05-03231.1995
14.
Tate S, Benn S, Hick C, Trezise D, John V, Mannion RJ, Costigan M, Plumpton C, Grose D, Gladwell Z, et al. 1998. Two sodium channels contribute to the TTX-R sodium current in primary sensory neurons. Nat Neurosci. 1(8):653-655. https://doi.org/10.1038/3652
15.
Toledo-Aral JJ, Moss BL, He Z, Koszowski AG, Whisenand T, Levinson SR, Wolf JJ, Silos-Santiago I, Halegoua S, Mandel G. 1997. Identification of PN1, a predominant voltage-dependent sodium channel expressed principally in peripheral neurons. Proceedings of the National Academy of Sciences. 94(4):1527-1532. https://doi.org/10.1073/pnas.94.4.1527
16.
Zakon HH. 2012. Adaptive evolution of voltage-gated sodium channels: The first 800 million years. Proceedings of the National Academy of Sciences. 109(Supplement_1):10619-10625. https://doi.org/10.1073/pnas.1201884109
17.
Zuliani V, Fantini M, Rivara M. 2012. Sodium Channel Blockers as Therapeutic Target for Treating Epilepsy: Recent Updates. CTMC. 12(9):962-970. https://doi.org/10.2174/156802612800229206