Determination of Water Content in 0.1 M Perchloric Acid in Acetic Acid Using Karl Fischer Titration
Product Group
Carboxylic acids, Inorganic compounds
General Information Concerning the Product Group
Carboxylic Acids
Most carboxylic acids can be analysed relatively easily following standard KF titration methods. Due to their weakly acidic properties, they exhibit only a slight effect on the pH of the KF solution. Only the stronger acids require neutralisation or buffering of the solvent prior to measuring. This is achieved by the use of organic amines. In some cases, esterification with methanol results in the formation of water. This potential interference should especially be taken into account with coulometric determination. In the event of solubility problems, solubilisers may be added. With some substances, only the adherent moisture can be determined by volumetric titration or, alternatively, KF oven technology may be employed.
Inorganic Compounds
Salts contain water in different kinds of bonding states: adherent moisture - enclosed water - crystal water. For determination of the total water content, the substance must be completely dissolved. If the salt does not dissolve in methanol sufficiently, formamide can be added to the recipient vessel and/or titration carried out at 50 °C. If adherent moisture alone is of interest, the dissolution of the sample must be prevented through addition of a non-polar solvent (e.g. chloroform) to the KF solvent. A good alternative for salts to the direct volumetric determination is often the Karl Fischer oven technique. Through slow heating of the sample, it is possible to differentiate among the various types of water bonding. For salts that cause disturbing side reactions in the KF solutions (e.g. carbonates, sulphites, borates), the KF oven technique is the method of choice. The same is also true for oxides and hydroxides. These alkaline compounds react with the weakly acidic KF solution, resulting in the formation of water, which is erroneously detected as water during the titration. Aqueous inorganic acids and bases are titrated volumetrically. Since stoichiometry and reaction rate of the Karl Fischer reaction are influenced by the pH, sufficient buffering of the KF solution is a prerequisite for reproducible and accurate results. Optimum conditions are obtained in the neutral pH range (pH 4 - 8). Thus, the determination of acids is performed through the addition of Aquastar® buffer solution for strong acids, imidazole or a derivative of imidazole to the titration media. For the titration of bases the pH is adjusted by addition of Aquastar® buffer solution for strong bases, salicylic acid or benzoic acid. With particularly strong, highly concentrated acids the propensity for esterification in methanolic solutions should also be noted. In such cases external neutralisation in a methanol-free medium is necessary.
Special Information Concerning the Sample and the Methods
Water determination in this mixture works in the same way as water determination in acetic acid. Both, the coulometric and the volumetric method can be used. Adequate buffering of the titration medium is necessary in order to give correct results. Consequently, when the volumetric method is used, the use of Aquastar® - Solvent or Aquastar® - CombiMethanol and Aquastar® - buffer solution for strong acids is recommended for one component titration. In coulometry the buffering capacity of the standard reagents is sufficient for approximately 7 g of sample per 100 mL of cell fill.
Titration One Component System
Reagents:
Titrant
188005 Aquastar® - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
Solvent
50 mL 188015 Aquastar® - Solvent - Solvent for volumetric titration with two component reagents
or
40 ml 188009 Aquastar® - CombiMethanol - and Aquastar® - buffer solution for strong acids Art.1.88035 10 ml
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
5 - 10 mL
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a plastic syringe (exact sample weight determination by weighing of syringe before and after injection) or volumetric pipette and the titration is started. In the case of strongly increasing cell drift and a significant increase in the titration time, the medium must be changed. Approximately 15 g of this sample can be determined without problems in one filling of titration medium.
Titration Two Component System
Reagents:
Titrant
188010 Aquastar® - Titrant 5 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 5 mg water
Solvent
50 mL 188015 Aquastar® - Solvent - Solvent for volumetric titration with two component reagents
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample Size:
5 - 10 mL
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a plastic syringe (exact sample weight determination by weighing of syringe before and after injection) or volumetric pipette and the titration is started. In the case of strongly increasing cell drift and a significant increase in the titration time, the medium must be changed. Approximately 15 g of this sample can be determined without problems in one filling of titration medium.
Coulometry with Diaphragm
Reagents:
Catholyte
5 mL 109255 Aquastar® - CombiCoulomat frit - Coulometric Karl Fischer reagent for cells with diaphragm
Anolyte
100 mL 109255 Aquastar® - CombiCoulomat frit - Coulometric Karl Fischer reagent for cells with diaphragm
Titration Parameters:
Default coulometer settings for cell with diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion: drift < 10 µg/min
Sample Size:
1 mL
Procedure:
The Karl-Fischer reagent is placed into the cathode and anode compartment of the titration cell with diaphragm. The coulometer is started and the solvent is titrated dry. After preliminary titration and stabilisation of drift the sample is injected into the titration cell with a plastic syringe (exact sample weight determination by weighing of syringe before and after injection) and the water determination is started. Approximately 7 g of sample can be determined without problems in one filling of titration medium. After that the medium must be changed due to the strongly increasing drift.
Coulometry without Diaphragm
Reagents:
Working Medium
100 mL 109257 Aquastar® - CombiCoulomat fritless - Coulometric Karl Fischer reagent for cells with or without diaphragm
Titration Parameters:
Default coulometer settings for cell without diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion: drift < 10 µg/min
Sample Size:
1 mL
Procedure:
The Karl-Fischer reagent is placed into the titration cell without diaphragm. The coulometer is started and the solvent is titrated dry. After preliminary titration and stabilisation of drift the sample is injected into the titration cell with a plastic syringe (exact sample weight determination by weighing of syringe before and after injection) and the water determination is started. Approximately 7 g of sample can be determined without problems in one filling of titration medium. After that the medium must be changed due to the strongly increasing drift.