HPLC Analysis of Vitamins, Fat Soluble (A and E) on Discovery® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] methanol: [B] water (95:5, A:B)
flow rate
2 mL/min
column temp.
30 °C
detector
UV, 290 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany