HPLC Analysis of Fluoxetine Enantiomers on Astec® CHIROBIOTIC® V2
![HPLC Analysis of Fluoxetine Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/397/598/0570eac1-77ed-4432-8d49-e2df7f80c4e2/800/0570eac1-77ed-4432-8d49-e2df7f80c4e2.jpg)
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
15 mM ammonium acetate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC