Merck
HPLC Analysis of Bile Acids on Ascentis® Express C18
HPLC Analysis of Bile Acids on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
standard
Product No.
Description
Pricing
Deoxycholic acid
≥98% (HPLC)
Cholic acid
from bovine and/or ovine, ≥98%
Lithocholic acid
≥95%
Chenodeoxycholic acid
CONDITIONS
column
Ascentis Express C18, 5 cm x 4.6 mm I.D., 2.7 μm particles (53826-U)
mobile phase
[A] 10 mM ammonium acetate, pH 4 (adjusted with acetic acid); [B] acetonitrile; (80:20, A:B)
gradient
20 to 100% B in 10 min
flow rate
1.0 mL/min
column temp.
35 °C
detector
ESI(-) in SIR mode
injection
5 μL
sample
10 μg/mL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany