Merck
GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX G-TA
GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
Mandelic acid
99%
(R)-(−)-Mandelic acid
98%
(R)-(−)-Mandelic acid
ReagentPlus®, ≥99%
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
140 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
inj. temp.
250 °C
sample
peak 1: S(+)-methyl mandelate
peak 2: R(-)-methyl mandelate
Description
Analysis Note
Elution order reversal on CHIRALDEX B-DP.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC