Merck
GC Analysis of 1-(α-Naphthyl)ethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX B-DA
GC Analysis of 1-(α-Naphthyl)ethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ B-DA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-DA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
1-(1-Naphthyl)ethylamine
98%
CONDITIONS
column
Astec CHIRALDEX B-DA, 30 m x 0.25 mm I.D., 0.12 μm (72023AST)
oven
160 °C
detector
FID, 250 °C
carrier gas
helium, 35 psi
sample
peak 1: R(+)-1-(α-naphthyl)ethylamine
peak 2: S(-)-1-(α-naphthyl)ethylamine
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC