GC Analysis of Class 2 Residual Solvents on OVI-G43
![GC Analysis of Class 2 Residual Solvents on OVI-G43 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/802/535/6bf75fef-7c65-4d7f-a752-9706056a6135/800/6bf75fef-7c65-4d7f-a752-9706056a6135.jpg)
CONDITIONS
column
OVI-G43, 30 m x 0.53 mm I.D., 3.0 μm (25396)
oven
35 °C (15 min), 5 °C/min to 200 °C
inj. temp.
225 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec @ 35 °C
injection
1 μL, 33:1 split
liner
single taper
sample
28 analytes, at concentrations indicated in DMSO