GC Analysis of cis/trans FAME Column Performance Mix on SP®-2560 per AOAC Method Ce 1h-05
CONDITIONS
column
SP-2560, 100 m x 0.25 mm I.D., 0.20 μm (24056)
oven
180 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 1 mL/min
injection
1 μL, 100:1 split
sample
cis/trans FAME Column Performance Mix (40495-U)
Description
Analysis Note
Complex mixtures of cis/trans FAME isomers are difficult to separate. Baseline resolution may not be possible for some samples due to variation in GC instrument designs and column efficiencies. As a result, AOAC Method Ce 1h-05 recommends monitoring changes in column performance with a known FAME mixture closely matching the range of fatty acids under investigation.
The resolution factor for Peak 1 and Peak 2 of this mix (40495-U) is a good indicator of colum performance. Using one of the resolution equations listed on the Data Sheet, the resolution factor can be calculated. Resolution factors approaching 1.5 (baseline resolution) are desired.
For additional information, request Data Sheet T708019.
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC