GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59
CONDITIONS
column
SLB-IL59, 20 m x 0.18 mm I.D., 0.14 μm (custom)
oven
65 ºC (0.5 min), 25 ºC/min to 300 ºC (20 min)
inj. temp.
265 °C
detector
MSD, interface at 300 ºC, m/z = 50-500
carrier gas
helium, 50 psi constant pressure
injection
1 μL splitless (1 min)
liner
4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
sample
each PAH at 10 μg/mL in methylene chloride
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany