Merck
  • A comparison of HLA-DR and -DQ allele and haplotype frequencies in Trinidadian populations of African, South Asian, and mixed ancestry.
A comparison of HLA-DR and -DQ allele and haplotype frequencies in Trinidadian populations of African, South Asian, and mixed ancestry.
Human immunology (2002-10-24)
Christine V F Carrington, Elli Kondeatis, D Dan Ramdath, Paul J Norman, Robert W Vaughan, Henry A F Stephens
ABSTRACT
Using polymerase chain reaction-sequence-specific primer (PCR-SSP) typing, this study determined the frequency of human leukocyte antigen (HLA) DR- and -DQ alleles and haplotypes in individuals of African (n = 75), South Asian (n = 98), and mixed (n = 102) ancestry from the Caribbean island of Trinidad. We detected 19 different haplotypes containing DRB3, 8 containing DRB4, 6 containing DRB5, and 6 different haplotypes without DRB3/4/5 genes. Twenty-nine haplotypes were identified in Africans, 24 in the South Asians, and 32 in the mixed group. We detected significant differences between the populations, principally at the DQA1 and DQB1 loci, although the allele frequency for DRB1*0901 was highest in the Africans (p(c) < 0.05). Trinidad African and mixed groups were generally more diverse than the South Asians and displayed a wider range of DRB1-DQB1 associations; DQB1*02 and DQB1*0301 each associated with five to six different DRB1 alleles in the Africans and mixed group but only two in South Asians. In the Africans and the mixed group, DQB1*04 was found with DRB1*0302 and DRB1*04, but only with DRB1*08 in the South Asians. Trinidad Africans revealed consistencies with populations in Western, Central, and Northern Africa, but differed considerably from individual populations on the African continent. Trinidad South Asians displayed similar allele frequencies and associations to other populations from Northern India.