Merck
  • Preclinical efficacy and biological effects of the oral proteasome inhibitor ixazomib in diffuse large B-cell lymphoma.
Preclinical efficacy and biological effects of the oral proteasome inhibitor ixazomib in diffuse large B-cell lymphoma.
Oncotarget (2018-02-09)
Wei Liu, Juan Chen, Archito T Tamayo, Changgeng Ruan, Li Li, Shouhao Zhou, Chan Shen, Ken H Young, Jason Westin, Richard E Davis, Shimin Hu, Leonard J Medeiros, Richard J Ford, Lan V Pham
ABSTRACT
Despite advances in deciphering the molecular pathogenesis of diffuse large B-cell lymphoma (DLBCL), patients with relapsed/refractory disease, particularly those with adverse genetic features (e.g., mutated p53 or double hit lymphoma (DHL)) have very poor prognoses, and effective therapies are lacking. In this study we examined the preclinical efficacy and associated biological effects of the first oral proteasome inhibitor, ixazomib, in DLBCL
Sigma-Aldrich
MISSION® esiRNA, targeting human CHEK2