Merck
  • A complementary role for tetraspanin superfamily member TSSC6 and ADP purinergic P2Y
A complementary role for tetraspanin superfamily member TSSC6 and ADP purinergic P2Y
Thrombosis research (2017-11-28)
Mohammed Makkawi, David Howells, Mark D Wright, Denise E Jackson
ABSTRACT
Tumor-suppressing subchromosomal transferable fragment cDNA 6 (TSSC6) expression is restricted to hematopoietic organs and tissues where it plays a role in hematopoietic-cell function. The ADP purinergic receptor P2Y
Sigma-Aldrich
N6-Methyl-2′-deoxyadenosine