Merck
  • AMPK activation counteracts cardiac hypertrophy by reducing O-GlcNAcylation.
AMPK activation counteracts cardiac hypertrophy by reducing O-GlcNAcylation.
Nature communications (2018-01-27)
Roselle Gélinas, Florence Mailleux, Justine Dontaine, Laurent Bultot, Bénédicte Demeulder, Audrey Ginion, Evangelos P Daskalopoulos, Hrag Esfahani, Emilie Dubois-Deruy, Benjamin Lauzier, Chantal Gauthier, Aaron K Olson, Bertrand Bouchard, Christine Des Rosiers, Benoit Viollet, Kei Sakamoto, Jean-Luc Balligand, Jean-Louis Vanoverschelde, Christophe Beauloye, Sandrine Horman, Luc Bertrand
ABSTRACT
AMP-activated protein kinase (AMPK) has been shown to inhibit cardiac hypertrophy. Here, we show that submaximal AMPK activation blocks cardiomyocyte hypertrophy without affecting downstream targets previously suggested to be involved, such as p70 ribosomal S6 protein kinase, calcineurin/nuclear factor of activated T cells (NFAT) and extracellular signal-regulated kinases. Instead, cardiomyocyte hypertrophy is accompanied by increased protein O-GlcNAcylation, which is reversed by AMPK activation. Decreasing O-GlcNAcylation by inhibitors of the glutamine:fructose-6-phosphate aminotransferase (GFAT), blocks cardiomyocyte hypertrophy, mimicking AMPK activation. Conversely, O-GlcNAcylation-inducing agents counteract the anti-hypertrophic effect of AMPK. In vivo, AMPK activation prevents myocardial hypertrophy and the concomitant rise of O-GlcNAcylation in wild-type but not in AMPKα2-deficient mice. Treatment of wild-type mice with O-GlcNAcylation-inducing agents reverses AMPK action. Finally, we demonstrate that AMPK inhibits O-GlcNAcylation by mainly controlling GFAT phosphorylation, thereby reducing O-GlcNAcylation of proteins such as troponin T. We conclude that AMPK activation prevents cardiac hypertrophy predominantly by inhibiting O-GlcNAcylation.
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)