- N-substituted methyl maleamates as larvicidal compounds against Aedes aegypti (Diptera: Culicidae).
N-substituted methyl maleamates as larvicidal compounds against Aedes aegypti (Diptera: Culicidae).
Severe human arboviral diseases can be transmitted by the mosquito Aedes aegypti (L.), including dengue, chikungunya, zika, and yellow fever. The use of larvicides in containers that can result as potential breeding places and cannot be eliminated is the main alternative in control programs. However, their continuous and widespread use caused an increase in insecticide-resistant populations of this mosquito. The aim of this study was to evaluate the effect of three N-substituted methyl maleamates as larvicides on Ae. aegypti, the N-propyl methyl maleamate (PMM), N-butyl methyl maleamate (BMM), and N-hexyl methyl maleamate (HMM). These compounds could have a different mode of action from those larvicides known so far. We evaluated the larva mortality after 1 and 24 h of exposure and we found that mortality was fast and occurs within the first 60 min. HMM was slightly more effective with LC