Merck
  • P2X7 receptor-pannexin 1 interaction mediates extracellular alpha-synuclein-induced ATP release in neuroblastoma SH-SY5Y cells.
P2X7 receptor-pannexin 1 interaction mediates extracellular alpha-synuclein-induced ATP release in neuroblastoma SH-SY5Y cells.
Purinergic signalling (2017-05-19)
Anna Wilkaniec, Magdalena Gąssowska, Grzegorz A Czapski, Magdalena Cieślik, Grzegorz Sulkowski, Agata Adamczyk
ABSTRACT
Abnormalities of alpha-synuclein (ASN), the main component of protein deposits (Lewy bodies), were observed in Parkinson's disease (PD), dementia with Lewy bodies, Alzheimer's disease, and other neurodegenerative disorders. These alterations include increase in the levels of soluble ASN oligomers in the extracellular space. Numerous works have identified several mechanisms of their toxicity, including stimulation of the microglial P2X7 receptor leading to oxidative stress. While the significant role of purinergic signaling-particularly, P2 family receptors-in neurodegenerative disorders is well known, the interaction of extracellular soluble ASN with neuronal purinergic receptors is yet to be studied. Therefore, in this study, we have investigated the effect of ASN on P2 purinergic receptors and ATP-dependent signaling. We used neuroblastoma SH-SY5Y cell line and rat synaptoneurosomes treated with exogenous soluble ASN. The experiments were performed using spectrofluorometric, radiochemical, and immunochemical methods. We found the following: (i) ASN-induced intracellular free calcium mobilization in neuronal cells and nerve endings depends on the activation of purinergic P2X7 receptors; (ii) activation of P2X7 receptors leads to pannexin 1 recruitment to form an active complex responsible for ATP release; and (iii) ASN greatly decreases the activity of extracellular ecto-ATPase responsible for ATP degradation. Thus, it is concluded that purinergic receptors might be putative pharmacological targets in the molecular mechanism of extracellular ASN toxicity. Interference with P2X7 signaling seems to be a promising strategy for the prevention or therapy of PD and other neurodegenerative disorders.
Sigma-Aldrich
Anti-P2X7 Purinergic Receptor antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Pyridoxal phosphate-6-azo(benzene-2,4-disulfonic acid) tetrasodium salt hydrate, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-Purinergic Receptor P2Y1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder