Merck
  • Electrical current generation in microbial electrolysis cells by hyperthermophilic archaea Ferroglobus placidus and Geoglobus ahangari.
Electrical current generation in microbial electrolysis cells by hyperthermophilic archaea Ferroglobus placidus and Geoglobus ahangari.
Bioelectrochemistry (Amsterdam, Netherlands) (2017-10-11)
Yasemin D Yilmazel, Xiuping Zhu, Kyoung-Yeol Kim, Dawn E Holmes, Bruce E Logan
ABSTRACT
Few microorganisms have been examined for current generation under thermophilic (40-65°C) or hyperthermophilic temperatures (≥80°C) in microbial electrochemical systems. Two iron-reducing archaea from the family Archaeoglobaceae, Ferroglobus placidus and Geoglobus ahangari, showed electro-active behavior leading to current generation at hyperthermophilic temperatures in single-chamber microbial electrolysis cells (MECs). A current density (j) of 0.68±0.11A/m
Sigma-Aldrich
Sodium fumarate dibasic, ≥99%