Merck
  • Directed transdifferentiation of Müller glial cells to photoreceptors using the sonic hedgehog signaling pathway agonist purmorphamine.
Directed transdifferentiation of Müller glial cells to photoreceptors using the sonic hedgehog signaling pathway agonist purmorphamine.
Molecular medicine reports (2017-10-07)
Dandan Gu, Songtao Wang, Shuai Zhang, Peng Zhang, Guomin Zhou
ABSTRACT
Specification of distinct cell types from Müller glial cells is key to the potential application of endogenous repair in retinal regeneration. Sonic hedgehog (SHH) has been established as a potent mitogen for rat Müller glial cells, which also induces Müller glial cells to dedifferentiate and adopt the phenotype of rod photoreceptors. The present study investigated the effects of purmorphamine, a small molecule that activates the SHH‑pathway, in the proliferation, dedifferentiation and transdifferentiation of Müller glial cells, as determined by several methods including immunofluorescence, polymerase chain reaction and western blotting. It was demonstrated that it may be able to replace SHH for the regeneration of retinal neurons. Purmorphamine was revealed to stimulate the proliferation of Müller glial cells by increasing the expression of cyclin D1 and cyclin D3. In addition, purmorphamine‑treated Müller glial cells were induced to dedifferentiate by inducing the expression of progenitor‑specific markers; subsequently differentiating into rod‑like photoreceptors. Intraocular injection of purmorphamine promoted the activation of Müller glial cells, and in turn, the production of rod‑like photoreceptors in acute damaged retina. These results suggested that the endogenous neurogenic capacity of retinal Müller glial cells may be enhanced by this small molecular agonist of the SHH signaling pathway.
Sigma-Aldrich
Monoclonal Anti-Thy 1.1 antibody produced in mouse, clone TN-26, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Syntaxin antibody produced in mouse, clone HPC-1, ascites fluid
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone V9, ascites fluid
Sigma-Aldrich
Anti-Glutamine Synthetase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Rhodopsin antibody produced in mouse, clone 1D4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, Biotin Conjugate, clone rat-401, from mouse, biotin conjugate
Sigma-Aldrich
Anti-Protein kinase C antibody, Mouse monoclonal, clone MC5, purified from hybridoma cell culture
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)