Merck
Investigating the Intracellular Behaviors of Liposomal Nanohybrids
Theranostics (2018-02-22)
Dan Zhu, Zhuyuan Wang, Shenfei Zong, Yizhi Zhang, Chen Chen, Ruohu Zhang, Binfeng Yun, Yiping Cui
ABSTRACT
The recent proposition to combine liposomes with nanoparticles presents great opportunities to develop multifunctional drug delivery platforms. Although impressive progress has been made, attempts to elucidate the role nanoparticles play in the integral nanohybrids are still rather limited. Here, using surface-enhanced Raman scattering (SERS) technique, we investigate the influence of metal nanoparticles on the liposomal properties, ranging from drug release to intracellular movement. Specifically, we prepared SERS-active nanohybrids by attaching metal nanoparticles to liposomes and employed SERS signals to explore the intracellular behavior of the nanohybrids. Once deposited on the cell membrane, the nanohybrids entered tumor cells
Sigma-Aldrich
2-Mercapto-4-methyl-5-thiazoleacetic acid, 98%
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)