Merck
  • DNA Vaccination by Electroporation Amplifies Broadly Cross-Restricted Public TCR Clonotypes Shared with HIV Controllers.
DNA Vaccination by Electroporation Amplifies Broadly Cross-Restricted Public TCR Clonotypes Shared with HIV Controllers.
Journal of immunology (Baltimore, Md. : 1950) (2017-10-11)
Madhura Mukhopadhyay, Moran Galperin, Mandar Patgaonkar, Sandhya Vasan, David D Ho, Alexandre Nouël, Mathieu Claireaux, Daniela Benati, Olivier Lambotte, Yaoxing Huang, Lisa A Chakrabarti
ABSTRACT
Rare patients who spontaneously control HIV replication provide a useful model to inform HIV vaccine development. HIV controllers develop particularly efficient antiviral CD4
Sigma-Aldrich
Human APP / Amyloid Beta A4 Protein ELISA Kit, for serum, plasma, cell culture supernatants and urine