- Magnesium uptake by connecting fluid-phase endocytosis to an intracellular inorganic cation filter.
Magnesium uptake by connecting fluid-phase endocytosis to an intracellular inorganic cation filter.
Nature communications (2017-12-02)
Sandra H Klompmaker, Kid Kohl, Nicolas Fasel, Andreas Mayer
PMID29192218
ABSTRACT
Cells acquire free metals through plasma membrane transporters. But, in natural settings, sequestering agents often render metals inaccessible to transporters, limiting metal bioavailability. Here we identify a pathway for metal acquisition, allowing cells to cope with this situation. Under limited bioavailability of Mg
MATERIALS