Merck
  • MEX3C interacts with adaptor-related protein complex 2 and involves in miR-451a exosomal sorting.
MEX3C interacts with adaptor-related protein complex 2 and involves in miR-451a exosomal sorting.
PloS one (2017-10-06)
Pin Lu, Huanhuan Li, Ning Li, Ravi N Singh, Colin E Bishop, Xiangxian Chen, Baisong Lu
ABSTRACT
Some RNA species, especially microRNAs, are non-randomly sorted into exosomes, but how selectivity of RNA exosomal sorting is achieved is unknown. We found that all three variants of RNA-binding ubiquitin E3 ligase (MEX3C)-MEX3C-1, MEX3C-2, and MEX3C-3 -interact with adaptor-related protein complex 2 (AP-2), a cargo adaptor in clathrin-mediated endocytosis. MEX3C's C-terminal RING finger domain and the hnRNP K homology (KH) domain shared by the three MEX3C variants are both necessary for MEX3C/AP-2 interaction. MEX3C associates with the endolysosomal compartment through an endocytosis-like process. siRNA-mediated inhibition of the MEX3C or AP-2 complex substantially decreased exosomal but not cellular microRNA miR-451a expression. Exosomal sorting is ceramide-dependent but not ESCRT-dependent in microRNA miR-451a. That RNA-binding protein associates with membrane trafficking machinery, and that its involvement in exosomal microRNA expression, suggest the existence of a mechanism for specific recruiting of RNA molecules to endosomes for subsequent exosomal sorting.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
GW4869, ≥90% (NMR)
Sigma-Aldrich
MISSION® esiRNA, targeting human AP2M1
Sigma-Aldrich
MISSION® esiRNA, targeting human HGS
Sigma-Aldrich
MISSION® esiRNA, targeting human MEX3C