Merck
  • Broadband-Emitting 2 D Hybrid Organic-Inorganic Perovskite Based on Cyclohexane-bis(methylamonium) Cation.
Broadband-Emitting 2 D Hybrid Organic-Inorganic Perovskite Based on Cyclohexane-bis(methylamonium) Cation.
ChemSusChem (2017-07-29)
Ishita Neogi, Annalisa Bruno, Damodaran Bahulayan, Teck Wee Goh, Biplab Ghosh, Rakesh Ganguly, Daniele Cortecchia, Tze Chien Sum, Cesare Soci, Nripan Mathews, Subodh Gautam Mhaisalkar
ABSTRACT
A new broadband-emitting 2 D hybrid organic-inorganic perovskite (CyBMA)PbBr
Sigma-Aldrich
1,3-Cyclohexanebis(methylamine), mixture of isomers, 98%