Merck
  • New σ-phases in the Nb-X-Ga and Nb-X-Al systems (X = Ru, Rh, Pd, Ir, Pt, and Au).
New σ-phases in the Nb-X-Ga and Nb-X-Al systems (X = Ru, Rh, Pd, Ir, Pt, and Au).
Dalton transactions (Cambridge, England : 2003) (2017-10-07)
Elizabeth M Carnicom, Weiwei Xie, Tomasz Klimczuk, R J Cava
ABSTRACT
Previously unreported σ-phases in the ternary niobium-based systems Nb-X-Ga for X = Ru, Rh, Pd, Ir, Pt, Au, and Nb-X-Al for X = Ir, and Pt are presented, prepared by arc-melting followed by annealing at 1000 °C for 1 week. The phases are primitive tetragonal (P4
Sigma-Aldrich
Gallium, 99.99% trace metals basis